Learn More
Dicumarol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 204120250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dicumarol | |
66-76-2 | |
White | |
99% | |
C19H12O6 | |
25 g | |
15, 3100 | |
Solubility in water: negligible. Other solubilities: practically insoluble in alcohols,ether,aceton,soluble in aq.alkaline solutions,pyridine and,similar org.bases,slightly soluble in benzene and chloroform | |
OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12 | |
336.30 | |
CHEBI:4513 | |
99% |
99% | |
290.0°C to 292.0°C | |
Authentic | |
Glass bottle | |
MFCD00006857 | |
19, 197 | |
dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl | |
DOBMPNYZJYQDGZ-UHFFFAOYSA-N | |
4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one | |
54676038 | |
336.29 | |
Fine Crystalline Powder |
Chemical Identifiers
66-76-2 | |
336.30 | |
DOBMPNYZJYQDGZ-UHFFFAOYSA-N | |
54676038 | |
4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one |
C19H12O6 | |
MFCD00006857 | |
dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl | |
CHEBI:4513 | |
OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately cal
GHS Signal Word: Danger
EINECSNumber : 200-632-9
RTECSNumber : GN7875000
TSCA : TSCA
RUO – Research Use Only