missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dicumarol, 99%, Thermo Scientific Chemicals

$205.78 - $205.78

Chemical Identifiers

CAS 66-76-2
Molecular Formula C19H12O6
Molecular Weight (g/mol) 336.30
MDL Number MFCD00006857
InChI Key DOBMPNYZJYQDGZ-UHFFFAOYSA-N
Synonym dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl
PubChem CID 54676038
ChEBI CHEBI:4513
IUPAC Name 4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one
SMILES OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC204120250
SDS View Documents
Thermo Scientific Chemicals
204120250
25 g Glass bottle
Each for $205.78
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

66-76-2
336.30
DOBMPNYZJYQDGZ-UHFFFAOYSA-N
54676038
4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one
C19H12O6
MFCD00006857
dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl
CHEBI:4513
OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12

Specifications

66-76-2
White
99%
C19H12O6
25 g
15, 3100
Solubility in water: negligible. Other solubilities: practically insoluble in alcohols,ether,aceton,soluble in aq.alkaline solutions,pyridine and,similar org.bases,slightly soluble in benzene and chloroform
OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12
336.30
CHEBI:4513
99%
Dicumarol, 99%
290.0°C to 292.0°C
Authentic
Glass bottle
MFCD00006857
19, 197
dicumarol, dicoumarol, bishydroxycoumarin, dicoumarin, melitoxin, antitrombosin, baracoumin, dicoumal, dicumarine, acadyl
DOBMPNYZJYQDGZ-UHFFFAOYSA-N
4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one
54676038
336.29
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately cal

GHS Signal Word: Danger

EINECSNumber : 200-632-9

RTECSNumber : GN7875000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only