missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexano-18-crown-6, mixture of isomers, 97%

CAS: 16069-36-6 | C20H36O6 | 372.502 g/mol

Supplier:  Thermo Scientific Chemicals A1534403

 View more versions of this product

Catalog No. AAA1534403


Only null left
Explore available promotions

Description

Description

Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

Solubility
Soluble in Toluene. Slightly soluble (13g/L, 26°C) in water.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with strong acids, strong oxidizing agents.
Specifications

Specifications

Dicyclohexano-18-crown-6
16069-36-6
>110°C (230°F)
C20H36O6
1 g
1130529
14,2602
Soluble in Toluene. Slightly soluble (13g/L,26°C) in water.
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2
85955
97%
mixture of isomers
46°C to 53°C
Odorless
MFCD00005099
UN2811
Hygroscopic
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
BBGKDYHZQOSNMU-UHFFFAOYSA-N
372.502
372.5

Chemical Identifiers

16069-36-6
372.502
BBGKDYHZQOSNMU-UHFFFAOYSA-N
85955
C20H36O6
MFCD00005099
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 240-216-4

RTECSNumber : HP5385000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only