Learn More
Dicyclohexano-18-crown-6, mixture of isomers, 97%, Thermo Scientific Chemicals
$58.81 - $602.64
Chemical Identifiers
CAS | 16069-36-6 |
---|---|
Molecular Formula | C20H36O6 |
Molecular Weight (g/mol) | 372.502 |
MDL Number | MFCD00005099 |
InChI Key | BBGKDYHZQOSNMU-UHFFFAOYSA-N |
Synonym | dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612 |
PubChem CID | 85955 |
SMILES | C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1534403
|
Thermo Scientific Chemicals
A1534403 |
1 g |
Each for $58.81
|
|
||||||
AAA1534409
|
Thermo Scientific Chemicals
A1534409 |
10 g |
Each for $295.84
|
|
||||||
AAA1534414
|
Thermo Scientific Chemicals
A1534414 |
25 g |
Each for $602.64
|
|
||||||
Description
Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.
Solubility
Soluble in Toluene. Slightly soluble (13g/L, 26°C) in water.
Notes
Hygroscopic. Store under dry inert gas. Incompatible with strong acids, strong oxidizing agents.
Chemical Identifiers
16069-36-6 | |
372.502 | |
BBGKDYHZQOSNMU-UHFFFAOYSA-N | |
85955 |
C20H36O6 | |
MFCD00005099 | |
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612 | |
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2 |
Specifications
16069-36-6 | |
>110°C (230°F) | |
C20H36O6 | |
1 g | |
1130529 | |
14,2602 | |
Soluble in Toluene. Slightly soluble (13g/L,26°C) in water. | |
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2 | |
85955 | |
97% |
46°C to 53°C | |
Odorless | |
MFCD00005099 | |
UN2811 | |
Hygroscopic | |
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612 | |
BBGKDYHZQOSNMU-UHFFFAOYSA-N | |
372.502 | |
372.5 | |
Dicyclohexano-18-crown-6, mixture of isomers |
Safety and Handling
GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 240-216-4
RTECSNumber : HP5385000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only