missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Didodecyldimethylammonium bromide, 99%, Thermo Scientific Chemicals

$288.17 - $288.17

Chemical Identifiers

CAS 3282-73-3
Molecular Formula C26H56BrN
Molecular Weight (g/mol) 462.64
MDL Number MFCD00041969
InChI Key XRWMGCFJVKDVMD-UHFFFAOYSA-M
Synonym didodecyldimethylammonium bromide, dilauryldimethylammonium bromide, n-dodecyl-n,n-dimethyldodecan-1-aminium bromide, didodecyl dimethyl ammonium bromide, unii-7y37a25k2i, dimethyldidodecylammonium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide, didodecyl dimethyl azanium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide 1:1
PubChem CID 18669
IUPAC Name didodecyl(dimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Br-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC407120250
SDS View Documents
Thermo Scientific Chemicals
407120250
25 g Plastic bottle
Each for $288.17
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

3282-73-3
462.64
XRWMGCFJVKDVMD-UHFFFAOYSA-M
18669
CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Br-]
C26H56BrN
MFCD00041969
didodecyldimethylammonium bromide, dilauryldimethylammonium bromide, n-dodecyl-n,n-dimethyldodecan-1-aminium bromide, didodecyl dimethyl ammonium bromide, unii-7y37a25k2i, dimethyldidodecylammonium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide, didodecyl dimethyl azanium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide 1:1
didodecyl(dimethyl)azanium;bromide

Specifications

3282-73-3
White
98.5% min. (Argentometry)
C26H56BrN
MFCD00041969
didodecyldimethylammonium bromide, dilauryldimethylammonium bromide, n-dodecyl-n,n-dimethyldodecan-1-aminium bromide, didodecyl dimethyl ammonium bromide, unii-7y37a25k2i, dimethyldidodecylammonium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide, didodecyl dimethyl azanium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide 1:1
CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Br-]
462.64
462.64
Crystalline Powder
161.0°C to 164.0°C
Authentic
Plastic bottle
(CH3(CH2)11)2N(Br)(CH3)2
25 g
XRWMGCFJVKDVMD-UHFFFAOYSA-M
didodecyl(dimethyl)azanium;bromide
18669
99%
Didodecyldimethylammonium bromide, 99%
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 221-923-7

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only