missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 2-(2-cyanoethyl)malonate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1156714

 View more versions of this product

Catalog No. AAA1156714


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethyl 2- (2-cyanoethyl)malonate
1.079
>110°C (230°F)
1.436
25 g
1790237
YJJLOESDBPRZIP-UHFFFAOYSA-N
diethyl 2-(2-cyanoethyl)propanedioate
87005
≥98%
17216-62-5
115°C to 117°C (2 mmHg)
C10H15NO4
MFCD00001966
UN3276
diethyl 2-2-cyanoethyl malonate, diethyl 2-cyanoethyl malonate, 1,3-diethyl 2-2-cyanoethyl propanedioate, diethyl2-2-cyanoethyl malonate, diethyl 2-2-cyanoethyl-malonate, propanedioic acid, 2-cyanoethyl-, diethyl ester, 2-cyanoethylmalonic acid diethylester, diethyl 2-2-cyanoethyl propane-1,3-dioate, pubchem14162
CCOC(=O)C(CCC#N)C(=O)OCC
213.233
213.23

Chemical Identifiers

17216-62-5
213.233
YJJLOESDBPRZIP-UHFFFAOYSA-N
diethyl 2-(2-cyanoethyl)propanedioate
C10H15NO4
MFCD00001966
87005
CCOC(=O)C(CCC#N)C(=O)OCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c

H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 241-260-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only