missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 2-(2-cyanoethyl)malonate, 98+%, Thermo Scientific Chemicals

$60.44 - $239.23

Chemical Identifiers

CAS 17216-62-5
Molecular Formula C10H15NO4
Molecular Weight (g/mol) 213.233
MDL Number MFCD00001966
InChI Key YJJLOESDBPRZIP-UHFFFAOYSA-N
PubChem CID 87005
IUPAC Name diethyl 2-(2-cyanoethyl)propanedioate
SMILES CCOC(=O)C(CCC#N)C(=O)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1156709
View Documents
Thermo Scientific Chemicals
A1156709
10 g
Each for $60.44
 
AAA1156714
View Documents
Thermo Scientific Chemicals
A1156714
25 g
Each for $143.73
 
AAA1156718
View Documents
Thermo Scientific Chemicals
A1156718
50 g
Each for $239.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17216-62-5
213.233
YJJLOESDBPRZIP-UHFFFAOYSA-N
diethyl 2-(2-cyanoethyl)propanedioate
C10H15NO4
MFCD00001966
87005
CCOC(=O)C(CCC#N)C(=O)OCC

Specifications

17216-62-5
115°C to 117°C (2 mmHg)
C10H15NO4
MFCD00001966
UN3276
diethyl 2-2-cyanoethyl malonate, diethyl 2-cyanoethyl malonate, 1,3-diethyl 2-2-cyanoethyl propanedioate, diethyl2-2-cyanoethyl malonate, diethyl 2-2-cyanoethyl-malonate, propanedioic acid, 2-cyanoethyl-, diethyl ester, 2-cyanoethylmalonic acid diethylester, diethyl 2-2-cyanoethyl propane-1,3-dioate, pubchem14162
CCOC(=O)C(CCC#N)C(=O)OCC
213.233
213.23
Diethyl 2- (2-cyanoethyl)malonate
1.079
>110°C (230°F)
1.436
10 g
1790237
YJJLOESDBPRZIP-UHFFFAOYSA-N
diethyl 2-(2-cyanoethyl)propanedioate
87005
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c

H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 241-260-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only