Learn More
Diethyl(3-pyridyl)borane, 98%, Thermo Scientific Chemicals
$88.21 - $1110.32
Chemical Identifiers
CAS | 89878-14-8 |
---|---|
Molecular Formula | C9H14BN |
Molecular Weight (g/mol) | 147.03 |
MDL Number | MFCD00012348 |
InChI Key | OJKBCQOJVMAHDX-UHFFFAOYSA-N |
Synonym | diethyl 3-pyridyl borane, 3-diethylboryl pyridine, diethyl 3-pyridinyl borane, pyridine, 3-diethylboryl, 3-diethylboranyl pyridine, diethyl-3-pyridylborane, dithyl 3-pyridyl borane, 3-pyridyldiethylborane, diethyl 3-pyridyl bo |
PubChem CID | 642851 |
IUPAC Name | diethyl(pyridin-3-yl)borane |
SMILES | CCB(CC)C1=CN=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6422506
|
Thermo Scientific Chemicals
H6422506 |
5 g |
Each for $88.21
|
|
|||||
AAH6422514
|
Thermo Scientific Chemicals
H6422514 |
25 g |
Each for $326.89
|
|
|||||
AAH6422522
|
Thermo Scientific Chemicals
H6422522 |
100 g |
Each for $1,110.32
|
|
|||||
Description
Reactant for: preparation of inhibitors of ataxia telangiectasia mutated and Rad3 related (ATR) protein kinase as potential anticancer agents, N-Methylation reactions of the pyridyl group, Palladium catalyzed amination reactions, intramolecular Heck reactions, Coupling reactions and Synthesis of trisubstituted pyrimidines from polyhalopyrimidines via Suzuki coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant for: preparation of inhibitors of ataxia telangiectasia mutated and Rad3 related (ATR) protein kinase as potential anticancer agents, N-Methylation reactions of the pyridyl group, Palladium catalyzed amination reactions, intramolecular Heck reactions, Coupling reactions and Synthesis of trisubstituted pyrimidines from polyhalopyrimidines via Suzuki coupling.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
89878-14-8 | |
147.03 | |
OJKBCQOJVMAHDX-UHFFFAOYSA-N | |
642851 | |
CCB(CC)C1=CN=CC=C1 |
C9H14BN | |
MFCD00012348 | |
diethyl 3-pyridyl borane, 3-diethylboryl pyridine, diethyl 3-pyridinyl borane, pyridine, 3-diethylboryl, 3-diethylboranyl pyridine, diethyl-3-pyridylborane, dithyl 3-pyridyl borane, 3-pyridyldiethylborane, diethyl 3-pyridyl bo | |
diethyl(pyridin-3-yl)borane |
Specifications
89878-14-8 | |
C9H14BN | |
5 g | |
diethyl 3-pyridyl borane, 3-diethylboryl pyridine, diethyl 3-pyridinyl borane, pyridine, 3-diethylboryl, 3-diethylboranyl pyridine, diethyl-3-pyridylborane, dithyl 3-pyridyl borane, 3-pyridyldiethylborane, diethyl 3-pyridyl bo | |
OJKBCQOJVMAHDX-UHFFFAOYSA-N | |
diethyl(pyridin-3-yl)borane | |
642851 | |
98% | |
Diethyl(3-pyridyl)borane |
172°C to 175°C (literature) | |
MFCD00012348 | |
4381494 | |
Slightly soluble in water. | |
CCB(CC)C1=CN=CC=C1 | |
147.03 | |
147.03 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only