missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 4-methylbenzylphosphonate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1519018

 View more versions of this product

Catalog No. AAA1519018


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethyl 4-methylbenzylphosphonate
1.07
>110°C (230°F)
1.496
50 g
diethyl 4-methylbenzylphosphonate, diethyl 4-methylbenzyl phosphonate, 4-methylbenzyl phosphonic acid diethyl ester, diethyl 4-methylphenyl methylphosphonate, diethoxy 4-methylphenyl methyl phosphino-1-one, acmc-209iuf, diethyl-4-methylbenzylphosphonate, 1-diethoxyphosphorylmethyl-4-methylbenzene, phosphonic acid, 4-methylphenyl methyl-, diethyl ester
CCOP(=O)(CC1=CC=C(C=C1)C)OCC
242.255
242.26
3762-25-2
146°C to 148°C (0.6 mmHg)
C12H19O3P
MFCD00041385
2940626
QKGBKPZAXXBLJE-UHFFFAOYSA-N
1-(diethoxyphosphorylmethyl)-4-methylbenzene
3050
≥98%

Chemical Identifiers

3762-25-2
242.255
QKGBKPZAXXBLJE-UHFFFAOYSA-N
3050
CCOP(=O)(CC1=CC=C(C=C1)C)OCC
C12H19O3P
MFCD00041385
diethyl 4-methylbenzylphosphonate, diethyl 4-methylbenzyl phosphonate, 4-methylbenzyl phosphonic acid diethyl ester, diethyl 4-methylphenyl methylphosphonate, diethoxy 4-methylphenyl methyl phosphino-1-one, acmc-209iuf, diethyl-4-methylbenzylphosphonate, 1-diethoxyphosphorylmethyl-4-methylbenzene, phosphonic acid, 4-methylphenyl methyl-, diethyl ester
1-(diethoxyphosphorylmethyl)-4-methylbenzene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-180-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only