missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 4-methylbenzylphosphonate, 98+%, Thermo Scientific Chemicals

$63.37 - $211.84

Chemical Identifiers

CAS 3762-25-2
Molecular Formula C12H19O3P
Molecular Weight (g/mol) 242.255
MDL Number MFCD00041385
InChI Key QKGBKPZAXXBLJE-UHFFFAOYSA-N
Synonym diethyl 4-methylbenzylphosphonate, diethyl 4-methylbenzyl phosphonate, 4-methylbenzyl phosphonic acid diethyl ester, diethyl 4-methylphenyl methylphosphonate, diethoxy 4-methylphenyl methyl phosphino-1-one, acmc-209iuf, diethyl-4-methylbenzylphosphonate, 1-diethoxyphosphorylmethyl-4-methylbenzene, phosphonic acid, 4-methylphenyl methyl-, diethyl ester
PubChem CID 3050
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-methylbenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)C)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1519009
View Documents
Thermo Scientific Chemicals
A1519009
10 g
Each for $63.37
 
AAA1519018
View Documents
Thermo Scientific Chemicals
A1519018
50 g
Each for $211.84
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3762-25-2
242.255
QKGBKPZAXXBLJE-UHFFFAOYSA-N
3050
CCOP(=O)(CC1=CC=C(C=C1)C)OCC
C12H19O3P
MFCD00041385
diethyl 4-methylbenzylphosphonate, diethyl 4-methylbenzyl phosphonate, 4-methylbenzyl phosphonic acid diethyl ester, diethyl 4-methylphenyl methylphosphonate, diethoxy 4-methylphenyl methyl phosphino-1-one, acmc-209iuf, diethyl-4-methylbenzylphosphonate, 1-diethoxyphosphorylmethyl-4-methylbenzene, phosphonic acid, 4-methylphenyl methyl-, diethyl ester
1-(diethoxyphosphorylmethyl)-4-methylbenzene

Specifications

3762-25-2
146°C to 148°C (0.6 mmHg)
C12H19O3P
MFCD00041385
2940626
QKGBKPZAXXBLJE-UHFFFAOYSA-N
1-(diethoxyphosphorylmethyl)-4-methylbenzene
3050
≥98%
1.07
>110°C (230°F)
1.496
10 g
diethyl 4-methylbenzylphosphonate, diethyl 4-methylbenzyl phosphonate, 4-methylbenzyl phosphonic acid diethyl ester, diethyl 4-methylphenyl methylphosphonate, diethoxy 4-methylphenyl methyl phosphino-1-one, acmc-209iuf, diethyl-4-methylbenzylphosphonate, 1-diethoxyphosphorylmethyl-4-methylbenzene, phosphonic acid, 4-methylphenyl methyl-, diethyl ester
CCOP(=O)(CC1=CC=C(C=C1)C)OCC
242.255
242.26
Diethyl 4-methylbenzylphosphonate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-180-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only