missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl acetylsuccinate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 113951000

 View more versions of this product

Catalog No. AC113951000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Diethyl acetylsuccinate
1.0800g/mL
>112°C
99%
C10H16O5
C2H5O2CCH2CH(COCH3)CO2C2H5
100g
1.08
Solubility in water: 15g/L (20°C)
CCOC(=O)CC(C(=O)C)C(=O)OCC
216.23
216.23
1115-30-6
180°C to 183°C (50.0mmHg)
Authentic
Glass bottle
1.4340 to 1.436
MFCD00009157
03, 801
diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
DVSDDICSXBCMQJ-UHFFFAOYSA-N
diethyl 2-acetylbutanedioate
66197
99%

Chemical Identifiers

1115-30-6
216.23
DVSDDICSXBCMQJ-UHFFFAOYSA-N
66197
CCOC(=O)CC(C(=O)C)C(=O)OCC
C10H16O5
MFCD00009157
diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
diethyl 2-acetylbutanedioate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-223-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only