missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl acetylsuccinate, 99%, Thermo Scientific™

Chemical Identifiers

CAS 1115-30-6
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.23
MDL Number MFCD00009157
InChI Key DVSDDICSXBCMQJ-UHFFFAOYSA-N
Synonym diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
PubChem CID 66197
IUPAC Name diethyl 2-acetylbutanedioate
SMILES CCOC(=O)CC(C(=O)C)C(=O)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC113950250
SDS View Documents
Thermo Scientific Chemicals
113950250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC113951000
SDS View Documents
Thermo Scientific Chemicals
113951000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC113955000
SDS View Documents
Thermo Scientific Chemicals
113955000
500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1115-30-6
216.23
DVSDDICSXBCMQJ-UHFFFAOYSA-N
66197
CCOC(=O)CC(C(=O)C)C(=O)OCC
C10H16O5
MFCD00009157
diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
diethyl 2-acetylbutanedioate

Specifications

1115-30-6
180°C to 183°C (50.0mmHg)
Authentic
Glass bottle
1.4340 to 1.436
MFCD00009157
03, 801
diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
DVSDDICSXBCMQJ-UHFFFAOYSA-N
diethyl 2-acetylbutanedioate
66197
99%
1.0800g/mL
>112°C
99%
C10H16O5
C2H5O2CCH2CH(COCH3)CO2C2H5
25g
1.08
Solubility in water: 15g/L (20°C)
CCOC(=O)CC(C(=O)C)C(=O)OCC
216.23
216.23
Diethyl acetylsuccinate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-223-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only