Learn More
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 364330010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl ether | |
Water <50 ppm, stabilized with BHT | |
Colorless | |
34.6°C | |
−45°C | |
Liquid | |
99.5% | |
C4H10O | |
MFCD00011646 | |
0.005% Max. (Coulometric) | |
11,3762 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12 | |
Extra Dry over Molecular Sieve |
-116.0°C | |
60-29-7 | |
0.7140g/mL | |
1 L | |
AcroSeal™ Glass Bottle | |
1.3510 to 1.3530 | |
0.714 | |
(CH3CH2)2O | |
0.2448 mPa.s (20°C) | |
01, 314 | |
15, 3861 | |
Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
99.5% |
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only