missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals

$70.69 - $382.96

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC364331000
SDS View Documents
Thermo Scientific Chemicals
364331000
100 mL AcroSeal™ Glass Bottle
Each for $70.69
 
AC364335000
SDS View Documents
Thermo Scientific Chemicals
364335000
500 mL Glass Bottle
Each for $123.09
 
AC364330010
SDS View Documents
Thermo Scientific Chemicals
364330010
1 L AcroSeal™ Glass Bottle
Each for $200.51
 
AC364330025
SDS View Documents
Thermo Scientific Chemicals
364330025
2.5 L AcroSeal™ Glass Bottle
Each for $382.96
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60-29-7
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC

Specifications

-116.0°C
Colorless
34.6°C
−45°C
Liquid
99.5%
C4H10O
MFCD00011646
0.005% Max. (Coulometric)
11,3762
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
Extra Dry over Molecular Sieve
60-29-7
0.7140g/mL
100 mL
AcroSeal™ Glass Bottle
1.3510 to 1.3530
0.714
(CH3CH2)2O
0.2448 mPa.s (20°C)
01, 314
15, 3861
Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils
CCOCC
74.12
CHEBI:35702
99.5%
Diethyl ether, Water <50 ppm, stabilized with BHT
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only