missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl ether, ACS reagent, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 615080040

 View more versions of this product

Catalog No. AC615080040



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethyl ether
60-29-7
34.6°C
-45°C (113°F)
0.001% max.
C4H10O
MFCD00011646
0.01% max.
11,3762
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.12
CHEBI:35702
≥99.0%
-116°C
0.7140g/mL
4 L
Pass Test
0.0002 meq/g max.
(CH3CH2)2O
Vapor Pressure: 587mbar at 20°C
01, 314
15, 3861
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
ACS Reagent

Chemical Identifiers

60-29-7
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

Emergency Overview
Extremely flammable liquid and vapor. Vapor may cause flash fire. May form explosive peroxides. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspir

Signal Word: DANGER!

EINECSNumber : 200-467-2

RTECSNumber : KI5775000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only