Learn More
Diethyl ether, ACS reagent, anhydrous, Thermo Scientific Chemicals
$116.33 - $1435.70
Chemical Identifiers
CAS | 60-29-7 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00011646 |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
IUPAC Name | ethoxyethane |
SMILES | CCOCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC615085000
|
Thermo Scientific Chemicals
615085000 |
500 mL |
Each for $116.33
|
|
|||||
AC615080010
|
Thermo Scientific Chemicals
615080010 |
1 L |
Each for $179.16
|
|
|||||
AC615080040
|
Thermo Scientific Chemicals
615080040 |
4 L |
Each for $571.12
Case of 4 Each for $0.00
|
|
|||||
AC615080200
|
Thermo Scientific Chemicals
615080200 |
20 L |
Each for $1,435.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Specifications
-116°C | |
0.7140g/mL | |
500 mL | |
Pass Test | |
0.001% max. | |
C4H10O | |
MFCD00011646 | |
0.01% max. | |
11,3762 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
≥99.0% | |
Diethyl ether |
60-29-7 | |
34.6°C | |
-45°C (113°F) | |
EcoSafPak™ Glass Bottle | |
0.0002 meq/g max. | |
(CH3CH2)2O | |
Vapor Pressure: 587mbar at 20°C | |
01, 314 | |
15, 3861 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12 | |
ACS Reagent |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Emergency Overview
Extremely flammable liquid and vapor. Vapor may cause flash fire. May form explosive peroxides. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspir
Signal Word: DANGER!
EINECSNumber : 200-467-2
RTECSNumber : KI5775000
TSCA : TSCA
RUO – Research Use Only