Learn More
Diethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1477209
Description
Diethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Diethyl methylphosphonate | |
1.04 | |
75°C (167°F) | |
1.414 | |
10 g | |
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h | |
NYYLZXREFNYPKB-UHFFFAOYSA-N | |
1-[ethoxy(methyl)phosphoryl]oxyethane | |
12685 | |
97% |
683-08-9 | |
194°C | |
C5H13O3P | |
MFCD00009813 | |
1753416 | |
Miscible with water. | |
CCOP(C)(=O)OCC | |
152.13 | |
152.13 |
Chemical Identifiers
683-08-9 | |
152.13 | |
NYYLZXREFNYPKB-UHFFFAOYSA-N | |
12685 | |
CCOP(C)(=O)OCC |
C5H13O3P | |
MFCD00009813 | |
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h | |
1-[ethoxy(methyl)phosphoryl]oxyethane |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 211-667-4
RTECSNumber : SZ9085000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only