Learn More
Diethyl methylphosphonate, 97%, Thermo Scientific Chemicals
$206.10 - $819.33
Chemical Identifiers
CAS | 683-08-9 |
---|---|
Molecular Formula | C5H13O3P |
Molecular Weight (g/mol) | 152.13 |
MDL Number | MFCD00009813 |
InChI Key | NYYLZXREFNYPKB-UHFFFAOYSA-N |
Synonym | diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h |
PubChem CID | 12685 |
IUPAC Name | 1-[ethoxy(methyl)phosphoryl]oxyethane |
SMILES | CCOP(C)(=O)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1477209
|
Thermo Scientific Chemicals
A1477209 |
10 g |
Each for $206.10
|
|
|||||
AAA1477214
|
Thermo Scientific Chemicals
A1477214 |
25 g |
Each for $414.11
|
|
|||||
AAA1477218
|
Thermo Scientific Chemicals
A1477218 |
50 g |
Each for $819.33
|
|
|||||
Description
Diethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
683-08-9 | |
152.13 | |
NYYLZXREFNYPKB-UHFFFAOYSA-N | |
12685 | |
CCOP(C)(=O)OCC |
C5H13O3P | |
MFCD00009813 | |
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h | |
1-[ethoxy(methyl)phosphoryl]oxyethane |
Specifications
683-08-9 | |
194°C | |
C5H13O3P | |
MFCD00009813 | |
1753416 | |
Miscible with water. | |
CCOP(C)(=O)OCC | |
152.13 | |
152.13 | |
Diethyl methylphosphonate |
1.04 | |
75°C (167°F) | |
1.414 | |
10 g | |
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h | |
NYYLZXREFNYPKB-UHFFFAOYSA-N | |
1-[ethoxy(methyl)phosphoryl]oxyethane | |
12685 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 211-667-4
RTECSNumber : SZ9085000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only