missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl phthalate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 114520010

 View more versions of this product

Catalog No. AC114520010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethyl phthalate
84-66-2
298.0°C to 299.0°C
Authentic
Glass bottle
1.5010 to 1.5030
1 L
1.118
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
FLKPEMZONWLCSK-UHFFFAOYSA-N
diethyl benzene-1,2-dicarboxylate
6781
CHEBI:34698
99%
99%
1.1180g/mL
156°C
98.5% min. (GC)
C12H14O4
MFCD00009111
09,798
15,7483
Solubility in water: 1g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,chloroform,carbon tetrachloride
CCOC(=O)C1=CC=CC=C1C(=O)OCC
222.24
12 mPa.s (20°C)
222.24
Liquid

Chemical Identifiers

84-66-2
222.24
FLKPEMZONWLCSK-UHFFFAOYSA-N
6781
diethyl benzene-1,2-dicarboxylate
C12H14O4
MFCD00009111
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
CHEBI:34698
CCOC(=O)C1=CC=CC=C1C(=O)OCC
Safety and Handling

Safety and Handling

EINECSNumber : 201-550-6

RTECSNumber : TI1050000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only