missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl phthalate, 99%, Thermo Scientific Chemicals

$38.21 - $599.70

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009111
InChI Key FLKPEMZONWLCSK-UHFFFAOYSA-N
Synonym diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
PubChem CID 6781
ChEBI CHEBI:34698
IUPAC Name diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)C1=CC=CC=C1C(=O)OCC
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC114520250
SDS View Documents
Thermo Scientific Chemicals
114520250
25 mL Glass bottle
Each for $38.21
 
AC114520010
SDS View Documents
Thermo Scientific Chemicals
114520010
1 L Glass bottle
Each for $112.97
 
AC114520025
SDS View Documents
Thermo Scientific Chemicals
114520025
2.5 L Glass bottle
Each for $236.16
 
AC114520100
SDS View Documents
Thermo Scientific Chemicals
114520100
10 L Plastic drum
Each for $599.70
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

84-66-2
222.24
FLKPEMZONWLCSK-UHFFFAOYSA-N
6781
diethyl benzene-1,2-dicarboxylate
C12H14O4
MFCD00009111
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
CHEBI:34698
CCOC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

84-66-2
298.0°C to 299.0°C
Authentic
Glass bottle
1.5010 to 1.5030
25 mL
1.118
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
FLKPEMZONWLCSK-UHFFFAOYSA-N
diethyl benzene-1,2-dicarboxylate
6781
CHEBI:34698
99%
Diethyl phthalate, 99%
1.1180g/mL
156°C
98.5% min. (GC)
C12H14O4
MFCD00009111
09,798
15,7483
Solubility in water: 1g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,chloroform,carbon tetrachloride
CCOC(=O)C1=CC=CC=C1C(=O)OCC
222.24
12 mPa.s (20°C)
222.24
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-550-6

RTECSNumber : TI1050000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only