missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl phthalate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 114520025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl phthalate | |
84-66-2 | |
298.0°C to 299.0°C | |
Authentic | |
Glass bottle | |
1.5010 to 1.5030 | |
2.5 L | |
1.118 | |
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e | |
FLKPEMZONWLCSK-UHFFFAOYSA-N | |
diethyl benzene-1,2-dicarboxylate | |
6781 | |
CHEBI:34698 | |
99% |
99% | |
1.1180g/mL | |
156°C | |
98.5% min. (GC) | |
C12H14O4 | |
MFCD00009111 | |
09,798 | |
15,7483 | |
Solubility in water: 1g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,chloroform,carbon tetrachloride | |
CCOC(=O)C1=CC=CC=C1C(=O)OCC | |
222.24 | |
12 mPa.s (20°C) | |
222.24 | |
Liquid |
Chemical Identifiers
84-66-2 | |
222.24 | |
FLKPEMZONWLCSK-UHFFFAOYSA-N | |
6781 | |
diethyl benzene-1,2-dicarboxylate |
C12H14O4 | |
MFCD00009111 | |
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e | |
CHEBI:34698 | |
CCOC(=O)C1=CC=CC=C1C(=O)OCC |
Safety and Handling
EINECSNumber : 201-550-6
RTECSNumber : TI1050000
TSCA : TSCA
RUO – Research Use Only