missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl succinate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 150540010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Diethyl succinate | |
| 0.2mg KOH/g max. | |
| 217.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.4190 to 1.4210 | |
| MFCD00009208 | |
| 02,609 | |
| 15,8999 | |
| Solubility in water: slightly soluble in water. Other solubilities: miscible with alcohol and ether | |
| CCOC(=O)CCC(=O)OCC | |
| 174.2 | |
| 174.2 |
| 123-25-1 | |
| 1.0400g/mL | |
| 90°C | |
| 97.5% min. (GC) | |
| C8H14O4 | |
| C2H5O2CCH2CH2CO2C2H5 | |
| 1kg | |
| 1.04 | |
| diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377 | |
| DKMROQRQHGEIOW-UHFFFAOYSA-N | |
| diethyl butanedioate | |
| 31249 | |
| 98% |
Chemical Identifiers
| 123-25-1 | |
| 174.2 | |
| DKMROQRQHGEIOW-UHFFFAOYSA-N | |
| 31249 | |
| CCOC(=O)CCC(=O)OCC |
| C8H14O4 | |
| MFCD00009208 | |
| diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377 | |
| diethyl butanedioate |
Safety and Handling
EINECSNumber : 204-612-
RUO â Research Use Only