missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl succinate, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 123-25-1 |
|---|---|
| Molecular Formula | C8H14O4 |
| Molecular Weight (g/mol) | 174.2 |
| MDL Number | MFCD00009208 |
| InChI Key | DKMROQRQHGEIOW-UHFFFAOYSA-N |
| Synonym | diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377 |
| PubChem CID | 31249 |
| IUPAC Name | diethyl butanedioate |
| SMILES | CCOC(=O)CCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC150540010
|
Thermo Scientific Chemicals
150540010 |
1kg | Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC150540051
|
Thermo Scientific Chemicals
150540051 |
5kg | Plastic drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC150540250
|
Thermo Scientific Chemicals
150540250 |
25kg | Plastic Can |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 123-25-1 | |
| 174.2 | |
| DKMROQRQHGEIOW-UHFFFAOYSA-N | |
| 31249 | |
| CCOC(=O)CCC(=O)OCC |
| C8H14O4 | |
| MFCD00009208 | |
| diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377 | |
| diethyl butanedioate |
Specifications
| 123-25-1 | |
| 1.0400g/mL | |
| 90°C | |
| 97.5% min. (GC) | |
| C8H14O4 | |
| C2H5O2CCH2CH2CO2C2H5 | |
| 1kg | |
| 1.04 | |
| diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377 | |
| DKMROQRQHGEIOW-UHFFFAOYSA-N | |
| diethyl butanedioate | |
| 31249 | |
| 98% |
| 0.2mg KOH/g max. | |
| 217.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.4190 to 1.4210 | |
| MFCD00009208 | |
| 02,609 | |
| 15,8999 | |
| Solubility in water: slightly soluble in water. Other solubilities: miscible with alcohol and ether | |
| CCOC(=O)CCC(=O)OCC | |
| 174.2 | |
| 174.2 | |
| Diethyl succinate |
Safety and Handling
EINECSNumber : 204-612-
RUO â Research Use Only