Learn More
Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
$45.13 - $122.03
Chemical Identifiers
CAS | 660-68-4 |
---|---|
MDL Number | MFCD00012499 |
InChI Key | HDITUCONWLWUJR-UHFFFAOYSA-N |
Synonym | diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl |
PubChem CID | 10197650 |
IUPAC Name | N-ethylethanamine;hydrochloride |
SMILES | CCNCC.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC155891000
|
Thermo Scientific Chemicals
155891000 |
100 g | Plastic bottle |
Each for $45.13
|
|
||||
AC155895000
|
Thermo Scientific Chemicals
155895000 |
500 g | Plastic bottle |
Each for $122.03
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
660-68-4 | |
HDITUCONWLWUJR-UHFFFAOYSA-N | |
10197650 | |
CCNCC.Cl |
MFCD00012499 | |
diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl | |
N-ethylethanamine;hydrochloride |
Specifications
660-68-4 | |
White | |
1% max. | |
99% | |
C4H11N·HCl | |
MFCD00012499 | |
04,95 | |
diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl | |
HDITUCONWLWUJR-UHFFFAOYSA-N | |
N-ethylethanamine;hydrochloride | |
10197650 | |
99% | |
Diethylamine hydrochloride, 99% |
227.0°C to 230.0°C | |
320.0°C to 330.0°C | |
Authentic | |
Plastic bottle | |
(C2H5)2NH·HCl | |
100 g | |
15,3122 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform,practically insoluble in ether | |
CCNCC.Cl | |
109.6 | |
109.6 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 211-541-9
RTECSNumber : IA3084000
TSCA : TSCA
RUO – Research Use Only