missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylenetriaminepentaacetic acid dianhydride, 95%, Thermo Scientific Chemicals

$94.79 - $306.79

Chemical Identifiers

CAS 23911-26-4
Molecular Formula C14H19N3O8
Molecular Weight (g/mol) 357.319
MDL Number MFCD00010697
InChI Key RAZLJUXJEOEYAM-UHFFFAOYSA-N
Synonym dtpa-anhydride, cyclic dtpa anhydride, 2-bis 2-2,6-dioxomorpholino ethyl amino acetic acid, unii-wnm9rlh5x9, wnm9rlh5x9, diethylenetriaminepentaacetic dianhydride, diethylenetriaminepentaacetic acid dianhydride, cdtpaa, glycine, n,n-bis 2-2,6-dioxo-4-morpholinyl ethyl, bis 2-2,6-dioxomorpholin-4-yl ethyl amino acetic acid
PubChem CID 100825
IUPAC Name 2-[bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino]acetic acid
SMILES C1C(=O)OC(=O)CN1CCN(CCN2CC(=O)OC(=O)C2)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1517803
View Documents
Thermo Scientific Chemicals
L1517803
1 g
Each for $94.79
 
AAL1517806
View Documents
Thermo Scientific Chemicals
L1517806
5 g
Each for $306.79
 
Description

Description

Diethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is-20°C. It is sensitive to moisture. Store under dry inert gas.
Specifications

Chemical Identifiers

23911-26-4
357.319
RAZLJUXJEOEYAM-UHFFFAOYSA-N
100825
C1C(=O)OC(=O)CN1CCN(CCN2CC(=O)OC(=O)C2)CC(=O)O
C14H19N3O8
MFCD00010697
dtpa-anhydride, cyclic dtpa anhydride, 2-bis 2-2,6-dioxomorpholino ethyl amino acetic acid, unii-wnm9rlh5x9, wnm9rlh5x9, diethylenetriaminepentaacetic dianhydride, diethylenetriaminepentaacetic acid dianhydride, cdtpaa, glycine, n,n-bis 2-2,6-dioxo-4-morpholinyl ethyl, bis 2-2,6-dioxomorpholin-4-yl ethyl amino acetic acid
2-[bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino]acetic acid

Specifications

23911-26-4
C14H19N3O8
1 g
Moisture sensitive
Hydrolyzes in water.
C1C(=O)OC(=O)CN1CCN(CCN2CC(=O)OC(=O)C2)CC(=O)O
357.319
357.32
Diethylenetriaminepentaacetic acid dianhydride
∽190°C (decomposition)
MFCD00010697
1230954
dtpa-anhydride, cyclic dtpa anhydride, 2-bis 2-2,6-dioxomorpholino ethyl amino acetic acid, unii-wnm9rlh5x9, wnm9rlh5x9, diethylenetriaminepentaacetic dianhydride, diethylenetriaminepentaacetic acid dianhydride, cdtpaa, glycine, n,n-bis 2-2,6-dioxo-4-morpholinyl ethyl, bis 2-2,6-dioxomorpholin-4-yl ethyl amino acetic acid
RAZLJUXJEOEYAM-UHFFFAOYSA-N
2-[bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino]acetic acid
100825
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only