missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylgermanium dichloride, Thermo Scientific Chemicals

As an intermediate

Supplier:  Thermo Scientific Chemicals 03410814

 View more versions of this product

Catalog No. AA3410814


Explore available promotions

Description

Description

As an intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethylgermanium dichloride
-37°C
1.372 g/mL
63°C (145°F)
C4H14Cl2Ge
MFCD00013589
UN3265
diethylgermanium dichloride, diethyldichlorogermane, diethyldichlorogermanium, dichloro diethyl germane, germane,dichlorodiethyl, acmc-1c1bs, diethylgermanium chloride, dichlorodiethylgermane
FVUWAWRMMUJVGO-UHFFFAOYSA-N
dichloro(diethyl)germane
83334
Liquid
13314-52-8
Colorless
175°C
Acrid
1.471
25 g
Moisture sensitive
Decomposes in water and NH3; Soluble in organic solvents
Cl.Cl.CC[GeH2]CC
205.69
201.64

Chemical Identifiers

13314-52-8
205.69
FVUWAWRMMUJVGO-UHFFFAOYSA-N
83334
Cl.Cl.CC[GeH2]CC
C4H14Cl2Ge
MFCD00013589
diethylgermanium dichloride, diethyldichlorogermane, diethyldichlorogermanium, dichloro diethyl germane, germane,dichlorodiethyl, acmc-1c1bs, diethylgermanium chloride, dichlorodiethylgermane
dichloro(diethyl)germane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 236-344-5

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only