missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylgermanium dichloride, Thermo Scientific Chemicals

As an intermediate

$814.48 - $2724.66

Chemical Identifiers

CAS 13314-52-8
Molecular Formula C4H14Cl2Ge
Molecular Weight (g/mol) 205.69
MDL Number MFCD00013589
InChI Key FVUWAWRMMUJVGO-UHFFFAOYSA-N
Synonym diethylgermanium dichloride, diethyldichlorogermane, diethyldichlorogermanium, dichloro diethyl germane, germane,dichlorodiethyl, acmc-1c1bs, diethylgermanium chloride, dichlorodiethylgermane
PubChem CID 83334
IUPAC Name dichloro(diethyl)germane
SMILES Cl.Cl.CC[GeH2]CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3410806
View Documents
Thermo Scientific Chemicals
03410806
5 g
Each for $814.48
 
AA3410814
View Documents
Thermo Scientific Chemicals
03410814
25 g
Each for $2,724.66
 
Description

Description

As an intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13314-52-8
205.69
FVUWAWRMMUJVGO-UHFFFAOYSA-N
83334
Cl.Cl.CC[GeH2]CC
C4H14Cl2Ge
MFCD00013589
diethylgermanium dichloride, diethyldichlorogermane, diethyldichlorogermanium, dichloro diethyl germane, germane,dichlorodiethyl, acmc-1c1bs, diethylgermanium chloride, dichlorodiethylgermane
dichloro(diethyl)germane

Specifications

13314-52-8
Colorless
175°C
Acrid
1.471
5 g
Moisture sensitive
Decomposes in water and NH3; Soluble in organic solvents
Cl.Cl.CC[GeH2]CC
205.69
201.64
Diethylgermanium dichloride
-37°C
1.372 g/mL
63°C (145°F)
C4H14Cl2Ge
MFCD00013589
UN3265
diethylgermanium dichloride, diethyldichlorogermane, diethyldichlorogermanium, dichloro diethyl germane, germane,dichlorodiethyl, acmc-1c1bs, diethylgermanium chloride, dichlorodiethylgermane
FVUWAWRMMUJVGO-UHFFFAOYSA-N
dichloro(diethyl)germane
83334
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 236-344-5

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only