missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Digital Heated Ultrasonic Cleaner

Easy to use with dependable digital control panel. The 40 kHz ultrasonic bath cleans a broad range of materials, and the tank is constructed of stainless steel to resist corrosion and is easy to clean.

Supplier:  Fisherbrand™ FUB9LD

 View more versions of this product

Catalog No. FB15337426


Explore available promotions

Description

Description

  • Programmable from 1 to 99 minutes
  • Stainless steel tank
  • Basket and cover included
  • 2-year warranty
  • Optional use of heating to 80°C to break down debris and oils
  • 40 kHz ultrasonic frequency
  • High and low cleaning power settings
  • Degas mode
Specifications

Specifications

10 A
Stainless Steel
40 kHz
9 L
Basket and cover
2 Years
1 to 99 min.
Digital
120 V
300 x 240 x 150 mm (11.8 x 9.4 x5.9 in.)
20°C to 80°C
Videos
SDS
Documents

Documents

Promotions

Promotions