missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diglycolic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2375814

 View more versions of this product

Catalog No. AAB2375814


Explore available promotions

Description

Description

Diglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions. It is also used in the synthesis of N-interlinked imipramines which show apoptotic activity against malignant cells as well as signs of combating Burkitt’s lymphoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions. It is also used in the synthesis of N-interlinked imipramines which show apoptotic activity against malignant cells as well as signs of combating Burkitt’s lymphoma.

Solubility
Soluble in water, methanol.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.
Specifications

Specifications

Diglycolic acid
110-99-6
MFCD00004309
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid
QEVGZEDELICMKH-UHFFFAOYSA-N
2-(carboxymethoxy)acetic acid
8088
98%
140°C to 145°C
C4H6O5
1072117
Soluble in water,methanol.
OC(=O)COCC(O)=O
134.09
134.09
25 g

Chemical Identifiers

110-99-6
134.09
QEVGZEDELICMKH-UHFFFAOYSA-N
8088
OC(=O)COCC(O)=O
C4H6O5
MFCD00004309
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid
2-(carboxymethoxy)acetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 203-823-5

RTECSNumber : AJ1530000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only