Learn More
Diglycolic acid, 98%, Thermo Scientific Chemicals
$65.21 - $529.59
Chemical Identifiers
CAS | 110-99-6 |
---|---|
Molecular Formula | C4H6O5 |
Molecular Weight (g/mol) | 134.09 |
MDL Number | MFCD00004309 |
InChI Key | QEVGZEDELICMKH-UHFFFAOYSA-N |
Synonym | diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid |
PubChem CID | 8088 |
IUPAC Name | 2-(carboxymethoxy)acetic acid |
SMILES | OC(=O)COCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2375814
|
Thermo Scientific Chemicals
B2375814 |
25 g |
Each for $65.21
|
|
|||||
AAB2375822
|
Thermo Scientific Chemicals
B2375822 |
100 g |
Each for $160.16
|
|
|||||
AAB2375836
|
Thermo Scientific Chemicals
B2375836 |
500 g |
Each for $529.59
|
|
|||||
Description
Diglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions. It is also used in the synthesis of N-interlinked imipramines which show apoptotic activity against malignant cells as well as signs of combating Burkittâs lymphoma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions. It is also used in the synthesis of N-interlinked imipramines which show apoptotic activity against malignant cells as well as signs of combating Burkittâs lymphoma.
Solubility
Soluble in water, methanol.
Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.
Chemical Identifiers
110-99-6 | |
134.09 | |
QEVGZEDELICMKH-UHFFFAOYSA-N | |
8088 | |
OC(=O)COCC(O)=O |
C4H6O5 | |
MFCD00004309 | |
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid | |
2-(carboxymethoxy)acetic acid |
Specifications
140°C to 145°C | |
C4H6O5 | |
1072117 | |
Soluble in water,methanol. | |
OC(=O)COCC(O)=O | |
134.09 | |
134.09 | |
25 g |
110-99-6 | |
MFCD00004309 | |
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid | |
QEVGZEDELICMKH-UHFFFAOYSA-N | |
2-(carboxymethoxy)acetic acid | |
8088 | |
98% | |
Diglycolic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 203-823-5
RTECSNumber : AJ1530000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only