missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dihydrogen hexafluorozirconate, 20% w/w aq. soln., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 039461A1

 View more versions of this product

Catalog No. AA39461A1


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dihydrogen hexafluorozirconate
Green
F6H2Zr
1 kg
PZFQYBOZXGWNAM-UHFFFAOYSA-H
207.22
Liquid
12021-95-3
1.52 g/mL
MFCD00082965
UN3264
207.23
20% w/w aq. soln.

Chemical Identifiers

12021-95-3
207.23
PZFQYBOZXGWNAM-UHFFFAOYSA-H
F6H2Zr
MFCD00082965
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H312-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302+H312-H314-H331-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 234-666-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only