Diisobutylaluminium hydride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Supplier:  Thermo Scientific Chemicals 183791000
 View more versions of this product
Catalog No. AC183791000
Explore available promotions
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagent
Specifications
Specifications
Diisobutylaluminium hydride
1191-15-7,110-54-3
0.7010g/mL
AcroSeal™ Glass bottle
[(CH3)2CHCH2]2AlH
100 mL
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
15, 3212
Solubility in water: reacts
142.22
1M Solution in Hexane
-70.0°C
−23°C
C8H19Al
MFCD00008928
04, IV, 4400
0.701
DIBAL-H
142.22
Solution