missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisobutylaluminium hydride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals

$108.90 - $731.77

Specifications

Chemical Name or Material Diisobutylaluminium hydride
Name Note 1M Solution in Hexane
CAS 1191-15-7,110-54-3
Melting Point -70.0°C
Density 0.7010g/mL
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC183791000
View Documents
Thermo Scientific Chemicals
183791000
100 mL AcroSeal™ Glass bottle
Each for $108.90
 
AC183794000
View Documents
Thermo Scientific Chemicals
183794000
400 mL AcroSeal™ Glass Bottle
Each for $387.52
 
AC183798000
View Documents
Thermo Scientific Chemicals
183798000
800 mL AcroSeal™ Glass Bottle
Each for $731.77
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid, Reduction reagent
Specifications

Specifications

Diisobutylaluminium hydride
1191-15-7,110-54-3
0.7010g/mL
C8H19Al
MFCD00008928
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
0.701
Solubility in water: reacts
142.22
1M Solution in Hexane
-70.0°C
−23°C
[(CH3)2CHCH2]2AlH
04, IV, 4400
15, 3212
DIBAL-H
142.22
Solution
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only