missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diisopropyl Adipate 99.0+%, TCI America™
Supplier: TCI America A0654500ML
Specifications
Diisopropyl Adipate | |
-1°C | |
120°C | |
MFCD00026391 | |
Adipic Acid Diisopropyl Ester | |
CC(C)OC(=O)CCCCC(=O)OC(C)C | |
230.30 | |
CHEBI:34711 | |
≥99.0% (GC) |
6938-94-9 | |
Yellow | |
C12H22O4 | |
500 mL | |
ZDQWESQEGGJUCH-UHFFFAOYSA-N | |
1,6-bis(propan-2-yl) hexanedioate | |
23368 | |
230.30 | |
Liquid |
Chemical Identifiers
6938-94-9 | |
230.30 | |
ZDQWESQEGGJUCH-UHFFFAOYSA-N | |
23368 | |
1,6-bis(propan-2-yl) hexanedioate |
C12H22O4 | |
MFCD00026391 | |
Adipic Acid Diisopropyl Ester | |
CHEBI:34711 | |
CC(C)OC(=O)CCCCC(=O)OC(C)C |
Safety and Handling
EINECSNumber : (2)-0861&(2)-0879
RTECSNumber : AV1575000
TSCA : Yes