missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl Adipate 99.0+%, TCI America™
$72.70 - $307.57
Chemical Identifiers
| CAS | 6938-94-9 |
|---|---|
| Molecular Formula | C12H22O4 |
| Molecular Weight (g/mol) | 230.30 |
| MDL Number | MFCD00026391 |
| InChI Key | ZDQWESQEGGJUCH-UHFFFAOYSA-N |
| Synonym | Adipic Acid Diisopropyl Ester |
| PubChem CID | 23368 |
| ChEBI | CHEBI:34711 |
| IUPAC Name | 1,6-bis(propan-2-yl) hexanedioate |
| SMILES | CC(C)OC(=O)CCCCC(=O)OC(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A065425ML
|
TCI America
A065425ML |
25 mL |
Each for $72.70
|
|
|||||
|
A0654500ML
|
TCI America
A0654500ML |
500 mL |
Each for $307.57
|
|
|||||
Chemical Identifiers
| 6938-94-9 | |
| 230.30 | |
| ZDQWESQEGGJUCH-UHFFFAOYSA-N | |
| 23368 | |
| 1,6-bis(propan-2-yl) hexanedioate |
| C12H22O4 | |
| MFCD00026391 | |
| Adipic Acid Diisopropyl Ester | |
| CHEBI:34711 | |
| CC(C)OC(=O)CCCCC(=O)OC(C)C |
Specifications
| 6938-94-9 | |
| Yellow | |
| C12H22O4 | |
| 25 mL | |
| ZDQWESQEGGJUCH-UHFFFAOYSA-N | |
| 1,6-bis(propan-2-yl) hexanedioate | |
| 23368 | |
| 230.30 | |
| Liquid |
| -1°C | |
| 120°C | |
| MFCD00026391 | |
| Adipic Acid Diisopropyl Ester | |
| CC(C)OC(=O)CCCCC(=O)OC(C)C | |
| 230.30 | |
| CHEBI:34711 | |
| ≥99.0% (GC) | |
| Diisopropyl Adipate |
Safety and Handling
EINECSNumber : (2)-0861&(2)-0879
RTECSNumber : AV1575000
TSCA : Yes