missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diisopropyl Trisulfide 98.0+%, TCI America™
Supplier: TCI America D430025G
Specifications
Diisopropyl Trisulfide | |
Yellow | |
C6H14S3 | |
25 g | |
CC(C)SSSC(C)C | |
182.36 | |
182.36 | |
Liquid |
5943-34-0 | |
79°C | |
MFCD00040042 | |
WUZSBMCRYUJOEU-UHFFFAOYSA-N | |
bis(propan-2-yl)trisulfane | |
80046 | |
≥98.0% (GC) |
Chemical Identifiers
5943-34-0 | |
182.36 | |
WUZSBMCRYUJOEU-UHFFFAOYSA-N | |
bis(propan-2-yl)trisulfane |
C6H14S3 | |
MFCD00040042 | |
80046 | |
CC(C)SSSC(C)C |
Safety and Handling
EINECSNumber : (2)-2421
TSCA : No