missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisopropyl Trisulfide 98.0+%, TCI America™

$205.89 - $622.84

Chemical Identifiers

CAS 5943-34-0
Molecular Formula C6H14S3
Molecular Weight (g/mol) 182.36
MDL Number MFCD00040042
InChI Key WUZSBMCRYUJOEU-UHFFFAOYSA-N
PubChem CID 80046
IUPAC Name bis(propan-2-yl)trisulfane
SMILES CC(C)SSSC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D43005G
View Documents
TCI America
D43005G
5 g
Each for $205.89
 
D430025G
View Documents
TCI America
D430025G
25 g
Each for $622.84
 
Description

Description

Specifications

Chemical Identifiers

5943-34-0
182.36
WUZSBMCRYUJOEU-UHFFFAOYSA-N
bis(propan-2-yl)trisulfane
C6H14S3
MFCD00040042
80046
CC(C)SSSC(C)C

Specifications

5943-34-0
79°C
MFCD00040042
WUZSBMCRYUJOEU-UHFFFAOYSA-N
bis(propan-2-yl)trisulfane
80046
≥98.0% (GC)
Diisopropyl Trisulfide
Yellow
C6H14S3
5 g
CC(C)SSSC(C)C
182.36
182.36
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2421

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'