missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisopropylamine, 99.5%, redistilled, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 378301000

 View more versions of this product

Catalog No. AC378301000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diisopropylamine
-61.0°C
84.0°C
Authentic
AcroSeal™ Glass Bottle
1.3910 to 1.3930
MFCD00008862
0.716
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
UAOMVDZJSHZZME-UHFFFAOYSA-N
N-propan-2-ylpropan-2-amine
7912
101.19
Liquid
108-18-9
0.7160g/mL
−17°C
99.4% min. (GC)
C6H15N
[(CH3)2CH]2NH
100 mL
15, 3215
Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol,soluble in most common organic solvents
CC(C)NC(C)C
101.19
0.4 mPa.s (20°C)
99.5%

Chemical Identifiers

108-18-9
101.19
UAOMVDZJSHZZME-UHFFFAOYSA-N
7912
CC(C)NC(C)C
C6H15N
MFCD00008862
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
N-propan-2-ylpropan-2-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

EINECSNumber : 203-558-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only