Learn More
Diisopropylamine, 99.5%, redistilled, AcroSeal™, Thermo Scientific Chemicals
$159.13 - $638.83
Chemical Identifiers
CAS | 108-18-9 |
---|---|
Molecular Formula | C6H15N |
Molecular Weight (g/mol) | 101.19 |
MDL Number | MFCD00008862 |
InChI Key | UAOMVDZJSHZZME-UHFFFAOYSA-N |
Synonym | diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine |
PubChem CID | 7912 |
IUPAC Name | N-propan-2-ylpropan-2-amine |
SMILES | CC(C)NC(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC378301000
|
Thermo Scientific Chemicals
378301000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $159.13
|
|
||||
AC378300010
|
Thermo Scientific Chemicals
378300010 |
1 L | AcroSeal™ Glass Bottle |
Each for $638.83
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-18-9 | |
101.19 | |
UAOMVDZJSHZZME-UHFFFAOYSA-N | |
7912 | |
CC(C)NC(C)C |
C6H15N | |
MFCD00008862 | |
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine | |
N-propan-2-ylpropan-2-amine |
Specifications
108-18-9 | |
0.7160g/mL | |
−17°C | |
99.4% min. (GC) | |
C6H15N | |
[(CH3)2CH]2NH | |
100 mL | |
15, 3215 | |
Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol,soluble in most common organic solvents | |
CC(C)NC(C)C | |
101.19 | |
0.4 mPa.s (20°C) | |
99.5% | |
Diisopropylamine |
-61.0°C | |
84.0°C | |
Authentic | |
AcroSeal™ Glass Bottle | |
1.3910 to 1.3930 | |
MFCD00008862 | |
0.716 | |
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine | |
UAOMVDZJSHZZME-UHFFFAOYSA-N | |
N-propan-2-ylpropan-2-amine | |
7912 | |
101.19 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Danger
EINECSNumber : 203-558-5
RUO – Research Use Only