Hardy Diagnostics™ Dilu-Lok™ Dilution Vials
Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries
Supplier:  Hardy Diagnostics™ D290
 View more versions of this product
Catalog No. 02-686-200A
Explore available promotions
Description
Description
Prefilled dilution vials with either 90 or 99mL of Butterfield's Buffer, Peptone Water (1%), or Phosphate Buffer.
  • Hinged lid provides leak-resistant, airtight seal and allows for easy one-handed opening
  • Tamper-evident shrink seal ensures sample integrity and are easily removed with perforated red tear strip
  • Flat surface on cap top ideal for marking sample identification
  • All diluents have a pH of 7.2 ±0.2
  • Wide 45mm opening accommodates larger samples and utensils
  • Made with optically clear polypropylene plastic, allowing for easy visualization of sample
  • Each lot meticulously tested for sterility, fill volume, pH, and toxicity
  • Certificates of Analysis are available
Compliance
Dilution vials meet APHA, AOAC, FDA, and USDA guidelines
Specifications
Specifications
APHA, AOAC, FDA, USDA
50 Cs.
Polypropylene
90 mL, Peptone Water 0.1%
Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries
Dilution Vial
Product Suggestions
Product Suggestions