missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hardy Diagnostics™ Dilu-Lok™ Dilution Vials
Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries
$345.74 - $1108.12
Specifications
Certifications/Compliance | APHA, AOAC, FDA, USDA |
---|---|
Quantity | 50 Cs. |
For Use With (Application) | Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries |
Material | Polypropylene |
Product Type | Dilution Vial |
Catalog Number | Mfr. No. | Includes | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | ||||||
02-686-200A
|
Hardy Diagnostics™
D290 |
90 mL, Peptone Water 0.1% |
Case of 50 Each for $1,108.12
|
|
||||||
02-686-200B
|
Hardy Diagnostics™
D299 |
99 mL, Peptone Water 0.1% |
Case of 50 Each for $1,092.62
|
|
||||||
02-686-201A
|
Hardy Diagnostics™
D590 |
90 mL, Butterfield's Phosphate Buffer |
Case of 50 Each for $353.62
|
|
||||||
02-686-202A
|
Hardy Diagnostics™
D690 |
90 mL, Phosphate Buffer w/ MgCl |
Case of 50 Each for $381.71
|
|
||||||
02-686-202B
|
Hardy Diagnostics™
D699 |
99 mL, Phosphate Buffer w/ MgCl |
Case of 50 Each for $392.06
|
|
||||||
02-686-201B
|
Hardy Diagnostics™
D599 |
99 mL, Butterfield's Phosphate Buffer |
Case of 50 Each for $345.74
|
|
||||||
Description
Prefilled dilution vials with either 90 or 99mL of Butterfield's Buffer, Peptone Water (1%), or Phosphate Buffer.
- Hinged lid provides leak-resistant, airtight seal and allows for easy one-handed opening
- Tamper-evident shrink seal ensures sample integrity and are easily removed with perforated red tear strip
- Flat surface on cap top ideal for marking sample identification
- All diluents have a pH of 7.2 ±0.2
- Wide 45mm opening accommodates larger samples and utensils
- Made with optically clear polypropylene plastic, allowing for easy visualization of sample
- Each lot meticulously tested for sterility, fill volume, pH, and toxicity
- Certificates of Analysis are available
Compliance
Dilution vials meet APHA, AOAC, FDA, and USDA guidelines
Specifications
APHA, AOAC, FDA, USDA | |
Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries | |
Dilution Vial |
50 Cs. | |
Polypropylene |