Hardy Diagnostics™ Dilu-Lok™ Dilution Vials
Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries
Supplier: Hardy Diagnostics™ D299
Description
Prefilled dilution vials with either 90 or 99mL of Butterfield's Buffer, Peptone Water (1%), or Phosphate Buffer.
- Hinged lid provides leak-resistant, airtight seal and allows for easy one-handed opening
- Tamper-evident shrink seal ensures sample integrity and are easily removed with perforated red tear strip
- Flat surface on cap top ideal for marking sample identification
- All diluents have a pH of 7.2 ±0.2
- Wide 45mm opening accommodates larger samples and utensils
- Made with optically clear polypropylene plastic, allowing for easy visualization of sample
- Each lot meticulously tested for sterility, fill volume, pH, and toxicity
- Certificates of Analysis are available
Compliance
Dilution vials meet APHA, AOAC, FDA, and USDA guidelines
Specifications
APHA, AOAC, FDA, USDA | |
50 Cs. | |
Polypropylene |
99 mL, Peptone Water 0.1% | |
Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries | |
Dilution Vial |