missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl allylphosphonate, tech. 85%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1056703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethyl allylphosphonate | |
41°C to 42°C (0.5 mmHg) | |
1.437 | |
1 g | |
dimethyl allylphosphonate, allyl phosphonic acid dimethyl ester, dimethyl prop-2-en-1-ylphosphonate, acmc-20al6z, dimethyl allyl phosphonate, allylphosphonic acid dimethyl, allylphosphonic acid dimethyl ester | |
COP(=O)(CC=C)OC | |
150.114 | |
150.12 | |
Technical |
757-54-0 | |
C5H11O3P | |
MFCD00014951 | |
1756526 | |
ZOSQAGGCVFVCNO-UHFFFAOYSA-N | |
3-dimethoxyphosphorylprop-1-ene | |
136570 | |
85% |
Chemical Identifiers
757-54-0 | |
150.114 | |
ZOSQAGGCVFVCNO-UHFFFAOYSA-N | |
136570 | |
COP(=O)(CC=C)OC |
C5H11O3P | |
MFCD00014951 | |
dimethyl allylphosphonate, allyl phosphonic acid dimethyl ester, dimethyl prop-2-en-1-ylphosphonate, acmc-20al6z, dimethyl allyl phosphonate, allylphosphonic acid dimethyl, allylphosphonic acid dimethyl ester | |
3-dimethoxyphosphorylprop-1-ene |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only