missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl glutarate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 116231000

 View more versions of this product

Catalog No. AC116231000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimethyl glutarate
1119-40-0
Colorless
210.0°C to 215.0°C
Authentic
Glass bottle
1.4236 to 1.4256
MFCD00008468
02, 633
15, 4509
Solubility in water: 53g/L
COC(=O)CCCC(=O)OC
160.17
2.5 mm2/s (25°C)
98%
98%
-37.0°C
1.0870g/mL
103°C
98%
C7H12O4
CH3O2C(CH2)3CO2CH3
100 g
1.087
dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester
XTDYIOOONNVFMA-UHFFFAOYSA-N
dimethyl pentanedioate
14242
160.17
Liquid

Chemical Identifiers

1119-40-0
160.17
XTDYIOOONNVFMA-UHFFFAOYSA-N
14242
COC(=O)CCCC(=O)OC
C7H12O4
MFCD00008468
dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester
dimethyl pentanedioate
Safety and Handling

Safety and Handling

EINECSNumber : 214-277-2

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only