missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl glutarate, 98%, Thermo Scientific Chemicals
$85.38 - $85.38
Chemical Identifiers
CAS | 1119-40-0 |
---|---|
Molecular Formula | C7H12O4 |
Molecular Weight (g/mol) | 160.17 |
MDL Number | MFCD00008468 |
InChI Key | XTDYIOOONNVFMA-UHFFFAOYSA-N |
Synonym | dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester |
PubChem CID | 14242 |
IUPAC Name | dimethyl pentanedioate |
SMILES | COC(=O)CCCC(=O)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC116231000
|
Thermo Scientific Chemicals
116231000 |
100 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1119-40-0 | |
160.17 | |
XTDYIOOONNVFMA-UHFFFAOYSA-N | |
14242 | |
COC(=O)CCCC(=O)OC |
C7H12O4 | |
MFCD00008468 | |
dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester | |
dimethyl pentanedioate |
Specifications
1119-40-0 | |
Colorless | |
210.0°C to 215.0°C | |
Authentic | |
Glass bottle | |
1.4236 to 1.4256 | |
MFCD00008468 | |
02, 633 | |
15, 4509 | |
Solubility in water: 53g/L | |
COC(=O)CCCC(=O)OC | |
160.17 | |
2.5 mm2/s (25°C) | |
98% | |
Dimethyl glutarate, 98% |
-37.0°C | |
1.0870g/mL | |
103°C | |
98% | |
C7H12O4 | |
CH3O2C(CH2)3CO2CH3 | |
100 g | |
1.087 | |
dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester | |
XTDYIOOONNVFMA-UHFFFAOYSA-N | |
dimethyl pentanedioate | |
14242 | |
160.17 | |
Liquid |
Safety and Handling
EINECSNumber : 214-277-2
TSCA : TSCA
RUO – Research Use Only