Learn More
Dimethyl oxalate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 159275000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Dimethyl oxalate | |
553-90-2 | |
163°C | |
Authentic | |
Plastic bottle | |
CH3O2CCO2CH3 | |
500g | |
1.14 | |
methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid | |
LOMVENUNSWAXEN-UHFFFAOYSA-N | |
dimethyl oxalate | |
11120 | |
118.09 |
99% | |
1.1400g/mL | |
75°C | |
98.5% min. (GC) | |
C4H6O4 | |
MFCD00008442 | |
02, 534 | |
15, 6179 | |
Solubility in water: 60g/L (25°C). Other solubilities: soluble in ethanol,ether,acetone,chloroform | |
COC(=O)C(=O)OC | |
118.09 | |
CHEBI:6859 | |
99% |
Chemical Identifiers
553-90-2 | |
118.09 | |
LOMVENUNSWAXEN-UHFFFAOYSA-N | |
11120 | |
dimethyl oxalate |
C4H6O4 | |
MFCD00008442 | |
methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid | |
CHEBI:6859 | |
COC(=O)C(=O)OC |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
EINECSNumber : 209-053-6
RTECSNumber : RO2850000
TSCA : TSCA
RUO â Research Use Only