missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl oxalate, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 553-90-2 |
---|---|
Molecular Formula | C4H6O4 |
Molecular Weight (g/mol) | 118.09 |
MDL Number | MFCD00008442 |
InChI Key | LOMVENUNSWAXEN-UHFFFAOYSA-N |
Synonym | methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid |
PubChem CID | 11120 |
ChEBI | CHEBI:6859 |
IUPAC Name | dimethyl oxalate |
SMILES | COC(=O)C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC159271000
|
Thermo Scientific Chemicals
159271000 |
100g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC159275000
|
Thermo Scientific Chemicals
159275000 |
500g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
553-90-2 | |
118.09 | |
LOMVENUNSWAXEN-UHFFFAOYSA-N | |
11120 | |
dimethyl oxalate |
C4H6O4 | |
MFCD00008442 | |
methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid | |
CHEBI:6859 | |
COC(=O)C(=O)OC |
Specifications
553-90-2 | |
163°C | |
Authentic | |
Plastic bottle | |
CH3O2CCO2CH3 | |
100g | |
1.14 | |
methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid | |
LOMVENUNSWAXEN-UHFFFAOYSA-N | |
dimethyl oxalate | |
11120 | |
118.09 | |
Dimethyl oxalate, 99% |
1.1400g/mL | |
75°C | |
98.5% min. (GC) | |
C4H6O4 | |
MFCD00008442 | |
02, 534 | |
15, 6179 | |
Solubility in water: 60g/L (25°C). Other solubilities: soluble in ethanol,ether,acetone,chloroform | |
COC(=O)C(=O)OC | |
118.09 | |
CHEBI:6859 | |
99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
EINECSNumber : 209-053-6
RTECSNumber : RO2850000
TSCA : TSCA
RUO â Research Use Only