missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl pyridine-2,5-dicarboxylate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1725006
Description
It is the intermediate in the synthesis of potent antitumor agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is the intermediate in the synthesis of potent antitumor agents.
Solubility
Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water.
Notes
Store at -20°C. Incompatible with oxidizing agents, reducing agents, bases.
Specifications
Dimethyl pyridine-2,5-dicarboxylate | |
160°C to 163°C | |
MFCD00034767 | |
172251 | |
Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water. | |
COC(=O)C1=CN=C(C=C1)C(=O)OC | |
195.174 | |
195.17 |
881-86-7 | |
C9H9NO4 | |
5 g | |
dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate | |
TUGSJNQAIMFEDY-UHFFFAOYSA-N | |
dimethyl pyridine-2,5-dicarboxylate | |
234911 | |
≥98% |
Chemical Identifiers
881-86-7 | |
195.174 | |
TUGSJNQAIMFEDY-UHFFFAOYSA-N | |
234911 | |
COC(=O)C1=CN=C(C=C1)C(=O)OC |
C9H9NO4 | |
MFCD00034767 | |
dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate | |
dimethyl pyridine-2,5-dicarboxylate |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only