missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl pyridine-2,5-dicarboxylate, 98+%, Thermo Scientific Chemicals

$75.04 - $936.84

Chemical Identifiers

CAS 881-86-7
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00034767
InChI Key TUGSJNQAIMFEDY-UHFFFAOYSA-N
Synonym dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate
PubChem CID 234911
IUPAC Name dimethyl pyridine-2,5-dicarboxylate
SMILES COC(=O)C1=CN=C(C=C1)C(=O)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1725006
View Documents
Thermo Scientific Chemicals
A1725006
5 g
Each for $75.04
 
AAA1725014
View Documents
Thermo Scientific Chemicals
A1725014
25 g
Each for $228.27
 
AAA1725022
View Documents
Thermo Scientific Chemicals
A1725022
100 g
Each for $936.84
 
Description

Description

It is the intermediate in the synthesis of potent antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the intermediate in the synthesis of potent antitumor agents.

Solubility
Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water.

Notes
Store at -20°C. Incompatible with oxidizing agents, reducing agents, bases.
Specifications

Chemical Identifiers

881-86-7
195.174
TUGSJNQAIMFEDY-UHFFFAOYSA-N
234911
COC(=O)C1=CN=C(C=C1)C(=O)OC
C9H9NO4
MFCD00034767
dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate
dimethyl pyridine-2,5-dicarboxylate

Specifications

881-86-7
C9H9NO4
5 g
dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate
TUGSJNQAIMFEDY-UHFFFAOYSA-N
dimethyl pyridine-2,5-dicarboxylate
234911
≥98%
160°C to 163°C
MFCD00034767
172251
Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water.
COC(=O)C1=CN=C(C=C1)C(=O)OC
195.174
195.17
Dimethyl pyridine-2,5-dicarboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only