missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl Sulfoxide, For headspace gas chromatography SupraSolv™ MilliporeSigma™

For headspace gas chromatography SupraSolv™

Supplier:  Technidata 1019001000

 View more versions of this product

Catalog No. M1019001000


Explore available promotions

Description

Description

Specifications

Specifications

Dimethyl Sulfoxide
67-68-5
1.10g/cm3 at 20°C
1 L
300°to302°C (572°to576°F)
Glass Bottle
1.48
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methanesulfinylmethane
679
78.13
18.5°C
Colorless
189°C
87°C (189°F)
Charactristic
Liquid
C2H6OS
0.6 hPa (20°C)
Soluble in water: 1000g/L at 20°C
CS(C)=O
78.13
CHEBI:28262

Chemical Identifiers

67-68-5
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
methanesulfinylmethane
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262
CS(C)=O
Safety and Handling

Safety and Handling

Recommended Storage : 15° to 25°C (59° to 77°F); Tightly closed. Dry

SDS
Documents

Documents

Promotions

Promotions